Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
This gene encodes a member of a highly conserved protein family that exists from archaea to vertebrates and plants. The encoded protein may function in RNA metabolism. Mutations within this gene are associated with Shwachman-Bodian-Diamond syndrome. An alternative transcript has been described, but its biological nature has not been determined. This gene has a closely linked pseudogene that is distally located.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10917; Ribosome maturation protein SBDS; Shwachman-Bodian-Diamond syndrome; Shwachman-Bodian-Diamond syndrome protein
Gene Aliases: CGI-97; SBDS; SDS; SWDS
UniProt ID: (Human) Q9Y3A5
Entrez Gene ID: (Human) 51119
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support