Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Positive control: Colon
Cellular location: Cytoplasm, mitochondria, nucleus.
SIRT3 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The SIRT3 is included in class I of the sirtuin family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:14931; hSIRT3; mitochondrial nicotinamide adenine dinucleotide-dependent deacetylase; mitochondrial protein lysine deacetylase; mSIR2L3; NAD-dependent deacetylase sirtuin-3; NAD-dependent deacetylase sirtuin-3, mitochondrial; NAD-dependent protein deacetylase sirtuin-3; NAD-dependent protein deacetylase sirtuin-3, mitochondrial; Regulatory protein SIR2 homolog 3; silent mating type information regulation 2, (S.cerevisiae, homolog)-like 3; silent mating type information regulation 2, S.cerevisiae, homolog 3; sir2-like 3; SIR2-like protein 3; SIRT3L mitochondrial; sirtuin type 3
Gene Aliases: 2310003L23Rik; AI848213; SIR2L3; SIRT3
UniProt ID: (Human) Q9NTG7, (Mouse) Q8R104
Entrez Gene ID: (Human) 23410, (Mouse) 64384, (Rat) 293615
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support