Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Quantity: 100 µg IgG per vial
Transcription factor that binds to the octamer motif (5'-ATTTGCAT-3'). Regulated the expression of a number of genes such as SPRR2A or placental lactogen.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epoc1; HGNC19864; Oct-11; Oct11; Octamer-binding protein 11; Octamer-binding transcription factor 11; OTF-11; OTF11; PLA1; POU domain transcription factor OCT11a; POU domain, class 2, transcription factor 3; POU2F3; Rov 1 pou-homeodomain protein; Transcription factor PLA-1; Transcription factor Skn-1
Gene Aliases: Epoc-1; OCT-11; OCT11; OTF-11; OTF11; PLA-1; PLA1; POU2F3; Skn-1; Skn-1a; Skn1
UniProt ID: (Human) Q9UKI9, (Rat) P42571
Entrez Gene ID: (Human) 25833, (Rat) 116544
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support