Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 7
Positive test controls include: HeLa, 293T, Mouse Liver. The target is usually found in the following locations: Cell membrane, Melanosome, Multi-pass membrane protein.
Immunogen sequence: MGKGVGRDKY EPAAVSEQGD KKGKKGKKDR DMDELKKEVS MDDHKLSLDE LHRKYGTDLS RGLTSARAAE ILARDGPNAL TPPPTTPEWI KFCRQLFGGF
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+/K+ transporting, alpha 4 polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 2; ATPase, Na+K+ transporting, alpha 1; Na(+)/K(+) ATPase alpha-1 subunit; Na(+)/K(+) ATPase alpha-4 subunit; Na+/K+ ATPase 1; Na+/K+ ATPase 4; Na+/K+ ATPase alpha 4 subunit; Na+/K+ ATPase, alpha-D polypeptide; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na,K-ATPase subunit alpha-C; sodium pump 4; Sodium pump subunit alpha-1; Sodium pump subunit alpha-4; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-4; sodium-potassium-ATPase, alpha 1 polypeptide; sodium/potassium-transporting ATPase alpha-4 chain; Sodium/potassium-transporting ATPase subunit alpha-1; Sodium/potassium-transporting ATPase subunit alpha-4
Gene Aliases: ATP1A1; ATP1A4; ATP1AL2; Atpa-1; BC010319; Nkaa1b
UniProt ID: (Human) P05023, (Human) Q13733, (Rat) P06685, (Mouse) Q8VDN2, (Mouse) Q9WV27, (Rat) Q64541
Entrez Gene ID: (Human) 476, (Human) 480, (Rat) 24211, (Mouse) 11928, (Mouse) 27222, (Rat) 29132
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support