Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Fabgennix
Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha and a beta subunit, catalyzes the conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GCS-alpha-2; guanylate cyclase 1, soluble, alpha 2; Guanylate cyclase soluble subunit alpha-2; soluble guanylyl cyclase alpha2 subunit
Gene Aliases: 6330407I18Rik; A230060L24Rik; GC-SA2; GUC1A2; GUCSA2; GUCY1A2
UniProt ID: (Human) P33402, (Rat) Q9WVI4
Entrez Gene ID: (Human) 2977, (Rat) 66012, (Mouse) 234889
Molecular Function:
guanylate cyclase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support