Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 8
Keep as concentrated solution.
Predicted reactivity: Mouse (100%), Rat (100%), Dog (100%), Pig (100%), Sheep (100%), Rhesus Monkey (100%), Bovine (100%).
Positive Control: adult mouse cerebellum.
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The hormone somatostatin has active 14 aa and 28 aa forms that are produced by alternate cleavage of the single preproprotein encoded by this gene. Somatostatin is expressed throughout the body and inhibits the release of numerous secondary hormones by binding to high-affinity G-protein-coupled somatostatin receptors. This hormone is an important regulator of the endocrine system through its interactions with pituitary growth hormone, thyroid stimulating hormone, and most hormones of the gastrointestinal tract. Somatostatin also affects rates of neurotransmission in the central nervous system and proliferation of both normal and tumorigenic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: preprosomatostatin; Somatostatin; somatostatin 28; somatotropin release-inhibiting factor; SRIF
Gene Aliases: Smst; SOM; SRIF; SS; SS-14; SS-28; Sst
UniProt ID: (Mouse) P60041, (Rat) P60042
Entrez Gene ID: (Mouse) 20604, (Rat) 24797
Molecular Function:
peptide hormone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support