Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
SSTR2 (somatostatin receptor 2) is a member of the superfamily of receptors having seven transmembrane segments and is expressed in highest levels in the cerebrum and kidney. Somatostatin acts at many sites to inhibit the release of many hormones and other secretory proteins. The receptor is coupled via pertussis toxin sensitive G proteins to inhibit adenylyl cyclase.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: somatostatin receptor; somatostatin receptor 2a; Somatostatin receptor type 2; SRIF-1; SS-2-R; SS2R; SSR2; SSTR2A
Gene Aliases: SSTR2
UniProt ID: (Human) P30874
Entrez Gene ID: (Human) 6752
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support