Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Immunogen sequence: RENDEFCKMG RYNLSPSIFF CATPPDDGNL CRFYKHNAAF CYKLPDNVTF EEGALIEPLS VGIHACRRGG VTLGHKVLVC GAGPIGMVTL LVAKAMGAAQ V
Sorbitol dehydrogenase catalyzes the interconversion of polyols and their corresponding ketoses, and together with aldose reductase is the reduction of glucose to sorbitol by ALDR1 with NADPH as the cofactor. SORD then oxidizes the sorbitol to fructose using NADcofactor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: (R,R)-butanediol dehydrogenase; epididymis secretory sperm binding protein Li 95n; L-iditol 2-dehydrogenase; Polyol dehydrogenase; RDH; Ribitol dehydrogenase; SDH; Sorbitol dehydrogenase; sorbitol dehydrogenase 1; SORD 1; XDH; Xylitol dehydrogenase
Gene Aliases: HEL-S-95n; Sdh-1; Sdh1; Sodh-1; SORD; SORD1
UniProt ID: (Human) Q00796, (Rat) P27867, (Mouse) Q64442
Entrez Gene ID: (Human) 6652, (Rat) 24788, (Mouse) 20322
Molecular Function:
dehydrogenase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support