Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
SOX17 (transcription factor SOX-17) belongs to the SRY-related HMB box family of transcription factors. It acts as a transcription regulator that binds target promoter DNA and bends the DNA. It binds to the sequences 5'-AACAAT-'3 or 5'-AACAAAG-3'. SOX17 modulates transcriptional regulation via WNT3A and inhibits Wnt signaling. It promotes degradation of activated CTNNB1. SOX17 plays an essential role in the regulation of embryonic development and is required for normal looping of the embryonic hear tube and for normal development of the definitive gut endoderm. Mutations in the gene can result in vesicoureteral reflux.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SRY (sex determining region Y)-box 17; SRY box 17; SRY-box 17; SRY-related HMG-box transcription factor SOX17; Transcription factor SOX-17
Gene Aliases: SOX17; VUR3
UniProt ID: (Human) Q9H6I2
Entrez Gene ID: (Human) 64321, (Dog) 486955
Molecular Function: HMG box transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support