Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the AAA (ATPases associated with a variety of cellular activities) protein family. Members of this protein family share an ATPase domain and have roles in diverse cellular processes including membrane trafficking, intracellular motility, organelle biogenesis, protein folding, and proteolysis. The encoded ATPase may be involved in the assembly or function of nuclear protein complexes. Two transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants have been described but their full length sequences have not been determined. Mutations associated with this gene cause the most frequent form of autosomal dominant spastic paraplegia 4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: fidgetin-like 1; KIAA183; SPAST; spastic paraplegia 4 (autosomal dominant; spastic paraplegia 4 (autosomal dominant; spastin); spastic paraplegia 4 homolog; Spastic paraplegia 4 protein; Spastin; spastin {ECO:0000255|HAMAP-Rule:MF_03021}
Gene Aliases: ADPSP; FSP2; KIAA1083; mKIAA1083; SPAST; SPG4
UniProt ID: (Human) Q9UBP0, (Mouse) Q9QYY8, (Rat) B2RYN7
Entrez Gene ID: (Human) 6683, (Mouse) 50850, (Rat) 362700
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support