Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
A 1:100 dilution of MA5-45723 was sufficient for detection of Beta 4 Spectrin in 20 µg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. |Detects approximately >200kDa. Does not cross react with other Beta-spectrins
This antibody was formerly sold as clone S393-2.
Spectrin is the major constituent of the cytoskeletal network underlying the erthrocyte plasma membrane and determines cell shape, arrangement of transmembrane proteins and organization of organelles. It is expressed at very low levels in many tissues, with the strongest expression in the cerebellum, spinal cord, stomach, pituitary gland, liver, pancreas, salivary gland, kidney, bladder and heart. Spectrin beta 4 is a non-erythrocyte member, and is expressed in the brain and pancreatic islets and localisted to the nuclear matrix, cytoplasmic vesicles and PML nuclear bodies. Beta4 spectrins are also essential for membrane stability and the molecular organization of the nodes of Ranvier.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beta-IV spectrin; beta-spectrin 4; EMBL BAB83243.1; lumbosacral neuroaxonal dystrophy; neuroaxonal dystrophy; quivering; spectrin beta 4; spectrin beta chain, brain 3; Spectrin beta chain, non-erythrocytic 4; spectrin, beta, non-erythrocytic 4; Spectrin, non-erythroid beta chain 3
Gene Aliases: 1700022P15Rik; 5830426A08Rik; dyn; KIAA1642; lnd; nmf261; nmf379; QV; ROSA62; SpbIV; SPNB4; SPTBN3; SPTBN4
UniProt ID: (Human) Q9H254
Entrez Gene ID: (Human) 57731, (Mouse) 80297, (Rat) 308458
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support