Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Medulla oblongata-derived protein; MGC17342; mob-1; Phosphatidylcholine:ceramide cholinephosphotransferase 1; Protein Mob; Sphingomyelin synthase 1; Transmembrane protein 23
Gene Aliases: 9530058O11Rik; AI841905; C80702; hmob33; MOB; MOB1; SGMS1; SMS1; Sor1; TMEM23
UniProt ID: (Human) Q86VZ5, (Mouse) Q8VCQ6, (Rat) Q7TSX5
Entrez Gene ID: (Human) 259230, (Mouse) 208449, (Rat) 353229
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support