Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive control: Human brain lysate (Cat. No. X1633C).
For Western blot, tissue lysate should be used at 10-50 µg/lane.
Sphingosine 1-phosphate (S1P) is a highly bioactive lipid that has a myriad of biological effects both intracellularly as a second messenger and extracellularly by binding to the S1P(1-5)/G-protein-coupled receptors of the endothelial differentiation gene family. Intracellularly, at least two enzymes, sphingosine kinase (1 and 2) and S1P phosphatase, regulate the activity of S1P by governing the phosphorylation of S1P. Regulation of hS1Pase1 with the resultant changes in cellular and secreted S1P has important implications in cell proliferation, angiogenesis and apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSPP1; hSPPase1; Sphingosine-1-phosphatase 1; Sphingosine-1-phosphate phosphatase 1; Sphingosine-1-phosphate phosphohydrolase 1; SPP-1; spp1; SPPase1
Gene Aliases: SGPP1; SPP1; SPPase1
UniProt ID: (Human) Q9BX95
Entrez Gene ID: (Human) 81537
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support