Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse
Transcription of protein-coding genes can be reconstituted on naked DNA with only the general transcription factors and RNA polymerase II. However, this minimal system cannot transcribe DNA packaged into chromatin, indicating that accessory factors may facilitate access to DNA. One such factor, FACT, interacts specifically with histones H2A/H2B to effect nucleosome disassembly and transcription elongation. FACT is composed of an 80 kDa subunit and a 140 kDa subunit; this gene encodes the 140 kDa subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Chromatin-specific transcription elongation factor 140 kDa subunit; facilitates chromatin remodeling 140 kDa subunit; Facilitates chromatin transcription complex subunit SPT16; FACT; FACT 140 kDa subunit; FACT complex subunit SPT16; FACTp140; FLJ10857; FLJ14010; FLJ34357; HGNC:11465; hSPT16; suppressor of Ty 16 homolog
Gene Aliases: CDC68; FACT140; FACTP140; SPT16; SPT16/CDC68; SUPT16H
UniProt ID: (Human) Q9Y5B9
Entrez Gene ID: (Human) 11198
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support