Invitrogen
MA1-83468 is specific for the Staphylococcus enterotoxins, and has shown the following reactivities when tested in ELISA:
enterotoxin B 100%, enterotoxin C1 18%, enterotoxin C2 82%, enterotoxin D 49%, enterotoxin E 24%.
Mouse anti Staphylococcus Enterotoxin A/B/C1/C2/D/E antibody, clone S13 Staphylococcus enterotoxin. It has shown the following reactivities when tested in ELISA. enterotoxin A65%enterotoxin B100%enterotoxin C118%enterotoxin C282%enterotoxin D49%enterotoxin E24%.