Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: LNKSEISQVF EIALKRNLPV NFEVARESGP PHMKNFVTKV SVGEFVGEGE GKSKKISKKN AAIAVLEELK KLPPLPAVER VKPRIKKKTK PIVKPQTSPE YGQGINPISR LAQIQQAKKE KEPEYTLLTE RGLPRRREFV MQVKVGNHTA EGTGTNKKVA KRNAAENMLE ILGFKVPQAQ
STAU1 is a member of a family of double-stranded RNA (dsRNA)-binding proteins involved in the transport and/or localization of mRNAs to different subcellular compartments and/or organelles. These proteins are characterized by the presence of multiple dsRNA-binding domains which are required to bind RNAs having double-stranded secondary structures. The human STAU1 also contains a microtubule- binding domain similar to that of microtubule-associated protein 1B, and binds tubulin. STAU1 has been shown to be present in the cytoplasm in association with the rough endoplasmic reticulum (RER), implicating this protein in the transport of mRNA via the microtubule network to the RER. STAU1 is also known to interact with influenza ribonucleoproteins NS1, NP, and PA and is required for efficient viral replication.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Double-stranded RNA-binding protein Staufen homolog 1; protein phosphatase 1, regulatory subunit 150; RP3-470L14.2; staufen RNA binding protein homolog 1; staufen, RNA binding protein, homolog 1
Gene Aliases: PPP1R150; STAU; STAU1
UniProt ID: (Human) O95793
Entrez Gene ID: (Human) 6780, (Rat) 84496
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support