Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thought to regulate cation conductance. May regulate ACCN1 and ACCN3 gating.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Erythrocyte band 7 integral membrane protein; Erythrocyte membrane protein band 7.2; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte protein band 7.2; erythrocyte protein band 7.2; protein 7.2b; erythrocyte surface protein band 7.2; Protein 7.2b; Stomatin
Gene Aliases: BND7; EPB7; Epb7.2; EPB72; STOM
UniProt ID: (Human) P27105, (Mouse) P54116
Entrez Gene ID: (Human) 2040, (Rat) 296655, (Mouse) 13830
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support