Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
FIGURE: 1 / 2
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EPB72-like 1; EPB72-like protein 1; Protein unc-24 homolog; SLP-1; stomatin (EBP72)-like 1; stomatin (EPB72)-like 1; stomatin-like 1; Stomatin-like protein 1; Stomatin-related protein; STORP
Gene Aliases: 1810015E19Rik; AI847431; hUNC-24; MSTP019; RGD1559463; SLP-1; SLP1; STOML1; STORP; UNC24; WPB72
UniProt ID: (Human) Q9UBI4, (Mouse) Q8CI66
Entrez Gene ID: (Human) 9399, (Rat) 300748, (Mouse) 69106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support