Search Thermo Fisher Scientific
FIGURE: 1 / 1
Reconstitute with 1 mL PBS.
Specificity: Strep II. A Step-tag is commonly added to recombinant proteins and can be used for detection or purification of the tagged protein.
Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ~ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.