Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Specificity: SuperNuclease (Recombinant Serratia marcescens Endonuclease)
SuperNuclease is a genetically-engineered endonuclease. It is a nonspecific nuclease with high activity, capable of completely digesting RNA and DNA (single-stranded, double-stranded, linear, circular and supercoiled forms, that no fewer than five phosphate residues) into 5'-monophosphate-terminated oligonucleotides of 3-5 bases in length. SuperNuclease can be removed by various purification methods.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA endonuclease; RNA endonuclease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support