NeoBiotechnologies
Positive Control:Human thymus
uterus or esophagus.
Cellular Location: Cytoplasm. Secreted.
SBSN is a protein coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HLAR698; Suprabasin
Gene Aliases: SBSN; UNQ698; UNQ698/PRO1343
UniProt ID: (Human) Q6UWP8
Entrez Gene ID: (Human) 374897
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support