Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 7
Synapsins (synapsin I, II, III) are a family of synaptic vesicle phosphoproteins that have been implicated in modulation of neurotransmitter release and in synaptogenesis. Synapsin II (SYN2) is composed of two isoforms, synapsin IIa and IIb.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: synapsin 2; Synapsin II; Synapsin-2; SYNIIa; SYNIIb
Gene Aliases: 2900074L19Rik; AI836018; AI841723; SYN2; SYNII
UniProt ID: (Human) Q92777, (Mouse) Q64332, (Rat) Q63537
Entrez Gene ID: (Human) 6854, (Mouse) 20965, (Rat) 29179
Molecular Function:
membrane trafficking regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support