Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene is a member of the synaptotagmin gene family and encodes a protein similar to other family members that mediate calcium-dependent regulation of membrane trafficking in synaptic transmission. Studies of the orthologous gene in rat have shown that the encoded protein selectively modulates spontaneous synaptic-vesicle exocytosis and may also be involved in regulating calcium independent secretion in nonneuronal cells. Alternative splicing results in multiple transcript variants. The gene has previously been referred to as synaptotagmin XI but has been renamed synaptotagmin XII to be standard with mouse and rat official nomenclature.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Srg1; synaptotagmin 12; Synaptotagmin XII; Synaptotagmin-12; Synaptotagmin-related gene 1 protein; synaptotagmin-XII; Synaptotagmin12; SynaptotagminXII; Syt XII; SYT-12; SytXII
Gene Aliases: Srg1; SYT11; SYT12; Sytr1; sytXII
UniProt ID: (Human) Q8IV01, (Mouse) Q920N7, (Rat) P97610
Entrez Gene ID: (Human) 91683, (Mouse) 171180, (Rat) 191595
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support