Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: AVPKNSLHL DLEKRDLNGN FPKTNLKPGS PSDLENATPK LFLEGEKESV SPESLKSSTS LTSEEKQEKL GTLFFSLEYN FERKAFVVNI KEARGLPAMD EQSMTSDPYI KMTILPEKKH KVKTRVLRKT LDPAFDETFT FYGIPYTQIQ ELALHFTILS FDRFSRDDII GEVLIPLSGI ELSEGKMLMN REIIKRNVRK SSGRGELLIS LCYQSTTNTL TVVVLKARHL PKSDVSGLSD PYVKVNLYHA KKRISKKKTH VKKCTPNAVF NELFVFDIPC EGLEDISVEF LVLDSERGSR NEVIGQLVLG AAAEGTGGEH WKEICDYPRR QIAKWHVLCD G (86-425 aa encoded by BC036538)
May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes (PubMed:23999003).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: synaptotagmin 4; Synaptotagmin IV; Synaptotagmin-4; Synaptotagmin4; SytIV
Gene Aliases: HsT1192; KIAA1342; Syt3; SYT4; SytIV
UniProt ID: (Human) Q9H2B2, (Rat) P50232, (Mouse) P40749
Entrez Gene ID: (Human) 6860, (Rat) 64440, (Mouse) 20983
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support