Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MATRRLTDA FLLLRNNSIQ NRQLLAEQLA DDRMALVSGI SLDPEAAIGV TKRPPPKWVD GVDEIQYDVG RIKQKMKELA SLHDKHLNRP TLDDSSEEEH AIEITTQEIT QLFHRCQRAV QALPSRARAC SEQEGRLLGN VVASLAQALQ ELSTSFRHAQ SGYLKRMKNR EERSQHFFDT SVPLMDDGDD NTLYHRGFTE DQLVLVEQNT LMVEEREREI RQIVQSISDL NEIFRDLGAM IVEQGTVLDR IDYNVEQSCI KTEDGLKQLH KAEQYQKKNR KMLVILILFV IIIVLIVVLV GVKSR (1-321 aa encoded by BC019042)
This gene encodes a protein that is a member of the syntaxin or t-SNARE (target-SNAP receptor) family. These proteins are found on cell membranes and serve as the targets for V-SNARES (vesicle-SNAP receptors) permitting specific synaptic vesicle docking and fusion. A microdeletion in the region of chromosome 20 where this gene is located has been associated with pseudohypoparathyroidism type Ib. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC90328; RP11-261P9.3; Syn16; Syntaxin-16
Gene Aliases: 4930401D03; 5430410K23Rik; 6330500A18Rik; AI648908; AW553605; STX16; SYN16
UniProt ID: (Human) O14662, (Mouse) Q8BVI5
Entrez Gene ID: (Human) 8675, (Mouse) 228960, (Rat) 362283
Molecular Function:
SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support