Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The gene is a member of the syntaxin family. The encoded protein is targeted to the apical membrane of epithelial cells where it forms clusters and is important in establishing and maintaining polarity necessary for protein trafficking involving vesicle fusion and exocytosis. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: syntaxin 3A; Syntaxin-3
Gene Aliases: STX3; STX3A; Syn-3
UniProt ID: (Human) Q13277, (Mouse) Q64704, (Rat) Q08849
Entrez Gene ID: (Human) 6809, (Mouse) 20908, (Rat) 81802
Molecular Function:
SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support