Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain, rat brain.
Predicted reactivity: Mouse (93%), Rat (94%), Pig (97%), Chicken (92%), Rhesus Monkey (99%), Bovine (97%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Syntaxin 6 is a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE domain, and an N-terminal regulatory domain. Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. Syntaxin is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HGNC:11441; Syn6; Syntaxin-6
Gene Aliases: 2310039E05Rik; 2410005I16Rik; AA437559; AI415714; AI845861; STX6
UniProt ID: (Human) O43752, (Mouse) Q9JKK1, (Rat) Q63635
Entrez Gene ID: (Human) 10228, (Mouse) 58244, (Rat) 60562
Molecular Function: SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support