Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSYTPGVGG DPAQLAQRIS SNIQKITQCS VEIQRTLNQL GTPQDSPELR QQLQQKQQYT NQLAKETDKY IKEFGSLPTT PSEQRQRKIQ KDRLVAEFTT SLTNFQKVQR QAAEREKEFV ARVRASSRVS GSFPEDSSKE RNLVSWESQT QPQVQVQDEE ITEDDLRLIH ERESSIRQLE ADIMDINEIF KDLGMMIHEQ GDVIDSIEAN VENAEVHVQQ ANQQLSRAAD YQRKSRKTLC IIILI (1-244 aa encoded by BC011975)
ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Syntaxin-7; Syntaxin7
Gene Aliases: AI315064; AI317144; AW107239; STX7; Syn7
UniProt ID: (Human) O15400, (Rat) O70257
Entrez Gene ID: (Human) 8417, (Rat) 60466, (Mouse) 53331
Molecular Function:
SNARE protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support