Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to TAC1.
Immunogen sequence: EEIGANDDL NYWSDWYDSD QIKEELPEPF EHLLQRIARR PKPQQFFGLM GKRDADSSIE KQVALLKALY GHGQISHKRH KTDSFVGLMG KRALNSVAYE RSAMQNYERR R (20-129 aa encoded by BC018047)
This gene encodes four products of the tachykinin peptide hormone family, substance P and neurokinin A, as well as the related peptides, neuropeptide K and neuropeptide gamma. These hormones are thought to function as neurotransmitters which interact with nerve receptors and smooth muscle cells. They are known to induce behavioral responses and function as vasodilators and secretagogues. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurokinin 1; neurokinin 2; neurokinin A; neurokinin alpha; neuromedin L; neuropeptide gamma; neuropeptide K; PPT; preprotachykinin; preprotachykinin A; protachykinin; Protachykinin-1; substance K; substance P; tachykinin 2; tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma)
Gene Aliases: 4930528L02Rik; Hs.2563; NK-1; NK1; NK2; NKA; NKNA; NPK; PPT-A; PPTA; SP; TAC1; TAC2
UniProt ID: (Human) P20366, (Mouse) P41539
Entrez Gene ID: (Human) 6863, (Mouse) 21333
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support