Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Predicted to react with Rabbit, Goat, Horse, Guinea pig, Dog, Pig, Rhesus monkey, Orangutan, Elephant (based on sequence homology)
Probably involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HF1L; SPT3-associated factor 42; SPT3-associated factor 42 (STAF42); STAF42; Transcriptional adapter 1; Transcriptional adapter 1-like protein; transcriptional adaptor 1 (HFI1 homolog, yeast)
Gene Aliases: ADA1; hADA1; HFI1; STAF42; TADA1; TADA1L
UniProt ID: (Human) Q96BN2
Entrez Gene ID: (Human) 117143
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support