Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLPPLIDPWD PGLTARDLLF RGGYRYRKRP RVVLDVTEQI SRFLLDHGDV AFAPLGKLML ENFKLEGAGS RTKKKTVVSV KKLLQDLGGH QPWGCPWAYL SNRQRRFSIL GGPILGTSVA SHLAELLHEE LVLRWEQLLL DEACTGGALA WVPGRTPQFG QLVYPAGGAQ DRLHFQEVVL TPGDNPQFLG KPGRIQLQGP VRQVVTCTVQ GETLLAVRSD YHCAVWKFGK QWQPTLLQAM QVEKGATGIS LSPHLPGELA ICSRSGAVCL WSPEDGLRQI YRDPETLVFR DSSSWRWADF TAHPRVLTVG DRTGVKMLDT QGPPGCGLLL FRLGAEASCQ KGERVLLTQY LGHSSPKCLP PTLHLVCTQF SLYLVDERLP LVPMLKWNHG LPSPLLLARL LPPPRPSCVQ PLLLGGQGGQ LQLLHLAEGA SVPRLAGPPQ SLPSRIDSLP AFPLLEPKIQ WRLQERLKAP TIGLAAVVPP MPSAPTPGLV LFQLSAAGDV FYQQLRPQVD SSLRRDAGPP GDTQPDCHAP TASWTSQDTA GCSQWLKALL KVPLAPPVWT APTFTHRQML GSTELRREEE EGQRLGVLRK AMARGQLLLQ RDLGSLPAAE PPPAPESGLE DKLSERLGEA WAGRGAAWWE RQQGRTSEPG RQTRRPKRRT QLSSSFSLSG HVDPSEDTSS PHSPEWPPAD ALPLPPTTPP SQELTPDACA QGVPSEQRQM LRDYMAKLPP QRDTPGCATT PPHSQASSVR ATRSQQHTPV LSSSQPLRKK PRMGF
Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. TAF1C is the largest SL1-specific TAF. Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase I-specific TBP-associated factor 110 kDa; SL1, 110kD subunit; TAFI110; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa; TATA box binding protein associated factor 1C; TATA box-binding protein-associated factor 1C; TATA box-binding protein-associated factor RNA polymerase I subunit C; TATA-box binding protein associated factor, RNA polymerase I, C; TBP-associated factor 1C; transcription factor SL1; Transcription initiation factor SL1/TIF-IB subunit C
Gene Aliases: MGC:39976; SL1; TAF1C; TAFI110; TAFI95
UniProt ID: (Human) Q15572
Entrez Gene ID: (Human) 9013
Molecular Function:
DNA-directed RNA polymerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support