Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: SLSGHVDPSE DTSSPHSPEW PPADALPLPP TTPPSQELTP DACAQGVPSE QRQMLRDYMA KLPPQRDTPG CATTPPHSQA SSVRATRSQQ HTPVLSSSQP LRKKPRMGF
Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. TAF1C is the largest SL1-specific TAF. Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the largest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase I-specific TBP-associated factor 110 kDa; SL1, 110kD subunit; TAFI110; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa; TATA box binding protein associated factor 1C; TATA box-binding protein-associated factor 1C; TATA box-binding protein-associated factor RNA polymerase I subunit C; TATA-box binding protein associated factor, RNA polymerase I, C; TBP-associated factor 1C; transcription factor SL1; Transcription initiation factor SL1/TIF-IB subunit C
Gene Aliases: MGC:39976; SL1; TAF1C; TAFI110; TAFI95
UniProt ID: (Human) Q15572
Entrez Gene ID: (Human) 9013
Molecular Function:
DNA-directed RNA polymerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support