Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes one of the larger subunits of TFIID that is stably associated with the TFIID complex. It contributes to interactions at and downstream of the transcription initiation site, interactions that help determine transcription complex response to activators.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 150 kDa cofactor of initiator function; cofactor of initiator function, 150kD subunit; RNA polymerase II TBP-associated factor subunit B; TAF(II)150; TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 150kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD; TATA box binding protein associated factor 2; TBP-associated factor 150 kDa; Transcription initiation factor TFIID 150 kDa subunit; Transcription initiation factor TFIID subunit 2
Gene Aliases: CIF150; MRT40; TAF2; TAF2B; TAFII150
UniProt ID: (Human) Q6P1X5
Entrez Gene ID: (Human) 6873
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support