Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MSEREERRFV EIPRESVRLM AESTGLELSD EVAALLAEDV CYRLREATQN SSQFMKHTKR RKLTVEDFNR ALRWSSVEAV CGYGSQEALP MRPAREGELY FPEDREVNLV ELALATNIPK GCAETAVRVH VSYLDGKGNL APQGSVPSAV SSLTDDLLKY YHQVTRAVLG DDPQLMKVAL QDLQTNSKIG ALLPYFVYVV SGVKSVSHDL EQLHRLLQVA RSLFRNPHLC LGPYVRCLVG SVLYCVLEPL AASINPLNDH WTLRDGAALL LSHIFWTHGD LVSGLYQHIL LSLQKILADP VRPLCCHYGA VVGLHALGWK AVERVLYPHL STYWTNLQAV LDDYSVSNAQ V
In eukaryotic systems, the process of initiating transcription from protein-coding genes requires the presence of RNA polymerase II and a broad family of auxiliary transcription factors. TFIID is a general transcription factor that initiates preinitiation complex assembly through direct interaction with the TATA promoter element. Functioning as a multisubunit complex consisting of a small TATA-binding polypeptide and other TBP-associated factors (TAFs), TFIID mediates promoter responses to various transcriptional activators and repressors. TAF6L, also known as PAF65A, is a 622 amino acid nuclear protein that functions as part of the PCAF (p300/CBP-associated factor) histone acetylase complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p300/CBP-associated factor (PCAF)-associated factor 65; PAF65-alpha; PCAF-associated factor 65 alpha; PCAF-associated factor 65-alpha; TAF6-like RNA polymerase II p300/CBP-associated factor-associated factor 65 kDa subunit 6L; TAF6-like RNA polymerase II, p300/CBP-associated factor (PCAF)-associated factor, 65kDa; TAF6L; TATA box binding protein associated factor 6 like
Gene Aliases: 2810417N14Rik; BB223262; C530024J06Rik; PAF65A; TAF6L
UniProt ID: (Human) Q9Y6J9, (Mouse) Q8R2K4
Entrez Gene ID: (Human) 10629, (Mouse) 225895, (Rat) 309194
Molecular Function:
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support