Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSKSKDDAP HELESQFILR LPPEYASTVR RAVQSGHVNL KDRLTIELHP DGRHGIVRVD RVPLASKLVD LPCVMESLKT IDKKTFYKTA DICQMLVSTV DGDLYPPVEE PVASTDPKAS KKKDKDKEKK FIWNHGITLP LKNVRKRRFR KTAKKKYIES PDVEKEVKRL LSTDAEAVST RWEIIAEDET KEAENQGLDI SSPGMSGHRQ GHDSLEHDEL REIFNDLSSS SEDEDETQHQ DEEDINIIDT EEDLERQLQD KLNESDEQHQ ENEGTNQLVM GIQKQIDNMK GKLQETQDRA KRQEDLIMKV ENLALKNRFQ AVLDELKQKE DREKEQLSSL QEELESLLEK (1-349 aa encoded by BC032737)
The intronless gene for this transcription coactivator is located between the protocadherin beta and gamma gene clusters on chromosome 5. The protein encoded by this gene is a component of the TFIID protein complex, a complex which binds to the TATA box in class II promoters and recruits RNA polymerase II and other factors. This particular subunit interacts with the largest TFIID subunit, as well as multiple transcription activators. The protein is required for transcription by promoters targeted by RNA polymerase II. The general transcription factor, TFIID, consists of the TATA-binding protein (TBP) associated with a series of TBP-associated factors (TAFs) that together participate in the assembly of the transcription preinitiation complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase II TBP-associated factor subunit F; TAF(II)55; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55 kDa; TAF7 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 55kDa; TAFII-55; TATA box binding protein (TBP) associated factor, RNA polymerase II, F, 55 kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, F, 55kD; TATA box binding protein associated factor 7; TBP-associated factor F; transcription factor IID subunit TAFII55; Transcription initiation factor TFIID 55 kDa subunit; Transcription initiation factor TFIID subunit 7; transcription initiation factor TFIID, 55 kDa subunit
Gene Aliases: 55kDa; AI607308; BB007175; TAF2F; TAF7; TAFII55
UniProt ID: (Human) Q15545, (Mouse) Q9R1C0
Entrez Gene ID: (Human) 6879, (Rat) 307485, (Mouse) 24074
Molecular Function:
RNA metabolism protein
nucleic acid metabolism protein
general transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support