Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MESGKMAPP KNAPRDALVM AQILKDMGIT EYEPRVINQM LEFAFRYVTT ILDDAKIYSS HAKKPNVDAD DVRLAIQCRA DQSFTSPPPR DFLLDIARQK NQTPLPLIKP YAGPRLPPDR YCLTAPNYRL KSLIKKGPNQ GRLVPRLSVG AVSSKPTTPT IATPQTVSVP NKVATPMSVT SQRFTVQIPP SQSTPVKPVP ATTAVQNVLI NPSMIGPKNI LITTNMVSSQ NTANEANPLK RKHEDDDDND IM (1-251 aa encoded by BC010350)
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DN-7; Neuronal cell death-related gene in neuron 7; neuronal cell death-related gene in neuron-7; Neuronal cell death-related protein 7; TAF9-like RNA polymerase II TATA box binding protein (TBP)-associated factor 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31 kD; TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, 31kDa; TATA box binding protein associated factor 9b; TBP-associated factor 9L; transcription associated factor TAFII31L; transcription initiation factor IID, 31kD subunit; Transcription initiation factor TFIID subunit 9-like; Transcription initiation factor TFIID subunit 9B; Transcription-associated factor TAFII31L
Gene Aliases: BC066223; DN-7; DN7; TAF9B; TAF9L; TAFII31; TAFII31L; TFIID-31
UniProt ID: (Human) Q9HBM6, (Mouse) Q6NZA9, (Rat) Q62880
Entrez Gene ID: (Human) 51616, (Mouse) 407786, (Rat) 171152
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support