Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Transgelin (also designated SM22alpha), is expressed abundantly in smooth muscle cells. Transgelin-2 (also known as SM22alpha homolog) is a homolog of transgelin and is also expressed in smooth muscle cells and by peritoneal B-1 cells. The human transgelin-2 gene (designated TAGLN2), which is located on chromosome 1q231-q25, encodes a 199 amino acid protein that contains a calponin-like repeat and a calponin-homology (CH) domain. Transgelin-2 may function very similarly to transgelin. During embryogenesis, transgelin is expressed in smooth, cardiac and skeletal muscle, but is restricted during late fetal development and adulthood to all vascular and visceral smooth muscle cells and low levels of expression in heart. Transgelin is downregulated in several transformed cell lines, indicating that a reduction of transgelin expression may be an early indicator of the onset of transformation. Transgelin also binds actin, causing actin fibers to gel within minutes of binding. Binding of transgelin to actin occurs at a ratio of 1:6 actin monomers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epididymis tissue protein Li 7e; SM22-alpha homolog; SM22-beta; Transgelin-2
Gene Aliases: 2700094C18Rik; CDABP0035; HA1756; KIAA0120; Sm22a; Sm22B; SM22beta; TAGLN2
UniProt ID: (Human) P37802, (Rat) Q5XFX0, (Mouse) Q9WVA4
Entrez Gene ID: (Human) 8407, (Rat) 304983, (Mouse) 21346
Molecular Function:
non-motor actin binding protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support