Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
TAIP2 binds to the consensus sequence 5'-AGAGTG-3' and has transcriptional activator activity. Plays a role in apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CSRNP-3; cysteine-serine-rich nuclear protein 3; Cysteine/serine-rich nuclear protein 3; family with sequence similarity 130, member A2; Protein FAM130A2; protein phosphatase 1, regulatory subunit 73; TAIP-2; TGF beta induced apotosis protein 2; TGF-beta induced apoptosis protein 2; TGF-beta-induced apoptosis protein 2
Gene Aliases: CSRNP3; FAM130A2; PPP1R73; TAIP-2; TAIP2
UniProt ID: (Human) Q8WYN3
Entrez Gene ID: (Human) 80034
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support