Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: IPTHPPDKKL SKNETLRLAM RYINFLVKVL GEQSLQQTGV AAQGNILGLF PQGPHLPGLE DRTLLENYQV PSPGPSHHIP
TAL2 is a helix-loop-helix protein. Translocations between this gene on chromosome 9 and the T-cell receptor beta-chain locus on chromosome 7 have been associated with activation of the T-cell acute lymphocytic leukemia 2 gene and T-cell acute lymphoblastic leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa19; Class A basic helix-loop-helix protein 19; T-cell acute lymphocytic leukemia 2; T-cell acute lymphocytic leukemia protein 2; TAL-2
Gene Aliases: BHLHA19; TAL2
UniProt ID: (Human) Q16559
Entrez Gene ID: (Human) 6887
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support