Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSSPVKRQ RMESALDQLK QFTTVVADTG DFHAIDEYKP QDATTNPSLI LAAAQMPAYQ ELVEEAIAYG RKLGGSQEDQ IKNAIDKLFV LFGAEILKKI PGRVSTEVDA RLSFDKDAMV ARARRLIELY KEAGISKDRI LIKLSSTWEG IQAGKELEEQ HGIHCNMTLL FSFAQAVACA EAGVTLISPF VGRILDWHVA NTDKKSYEPL EDPGVKSVTK IYNYYKKFSY KTIVMGASFR NTGEIKALAG CDFLTISPKL LGELLQDNAK LVPVLSAK (1-277 aa encoded by BC009680)
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dihydroxyacetone transferase; glycerone transferase; testicular secretory protein Li 56; Transaldolase
Gene Aliases: TAL; TAL-H; TALDO; TALDO1; TALDOR; TALH
UniProt ID: (Human) P37837, (Rat) Q9EQS0, (Mouse) Q93092
Entrez Gene ID: (Human) 6888, (Rat) 83688, (Mouse) 21351
Molecular Function:
aldolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support