Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVQATTLNV AFNSHNKSLL TIMMSNNFVE IKGSVFKKFE KNNLFQMSNS DIKERFTNYV LLLIVCLRNM EQFSWNPDHL WVLFPDVCMV IASEIAVDIV KHAFITKFND ITADVYSEYR ASLAFDLVSS RQKNAYTDYS DSVARRM (257-402 aa encoded by BC066899 )
This gene encodes a highly conserved, putative transmembrane protein. A mutation in the mouse ortholog of this gene results in homeotic, posterior-to-anterior transformations of the axial skeleton which are similar to the phenotype of mouse homeobox C8 gene mutants. This gene is proposed to function downstream of homeobox C8 to transduce extracellular patterning information during axial skeleton development. An alternatively spliced transcript variant encoding a substantially different isoform has been described, but its biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytomegalovirus partial fusion receptor; Transmembrane anterior posterior transformation protein 1; Transmembrane anterior posterior transformation protein 1 homolog
Gene Aliases: 4932414K18Rik; CMVFR; OCLSBG; TAPT1
UniProt ID: (Human) Q6NXT6, (Mouse) Q4VBD2
Entrez Gene ID: (Human) 202018, (Mouse) 231225
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support