Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Adding 0.2 mL of distilled water will yield a concentration of 500 µg/mL.
This gene encodes the L protein of the mitochondrial glycine cleavage system. The L protein, also named dihydrolipoamide dehydrogenase, is also a component of the pyruvate dehydrogenase complex, the alpha-ketoglutarate dehydrogenase complex, and the branched-chain alpha-keto acide dehydrogenase complex. Mutations in this gene have been identified in patients with E3-deficient maple syrup urine disease and lipoamide dehydrogenase deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Threonine--tRNA ligase, mitochondrial; Threonyl-tRNA synthetase; threonyl-tRNA synthetase, mitochondrial; Threonyl-tRNA synthetase-like 1; ThrRS
Gene Aliases: 2610024N01Rik; AI429208; COXPD21; RGD1308283; TARS2; TARSL1; thrRS
UniProt ID: (Human) Q9BW92, (Mouse) Q3UQ84, (Rat) Q68FW7
Entrez Gene ID: (Human) 80222, (Mouse) 71807, (Rat) 310672
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support