Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: TAS2R30.
Receptor that may play a role in the perception of bitterness and is gustducin-linked. May play a role in sensing the chemical composition of the gastrointestinal content. The activity of this receptor may stimulate alpha gustducin, mediate PLC-beta-2 activation and lead to the gating of TRPM5. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T2R30; T2R47; TAS2R44; taste receptor T2R47; Taste receptor type 2 member 30; Taste receptor type 2 member 47; taste receptor, type 2, member 30; taste receptor, type 2, member 47; type 2 taste receptor member 30; type 2 taste receptor member 47
Gene Aliases: T2R30; T2R47; TAS2R30; TAS2R47
UniProt ID: (Human) P59541
Entrez Gene ID: (Human) 259293
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support