Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total TAS2R40.
Gustducin-coupled receptor implicated in the perception of bitter compounds in the oral cavity and the gastrointestinal tract. Signals through PLCB2 and the calcium-regulated cation channel TRPM5. Expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells. Several bitter taste receptors are expressed in a single taste receptor cell.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 60; G-protein coupled receptor 60; MGC126844; MGC126846; T2R18; T2R40; T2R58; tas2r40 {ECO:0000250|UniProtKB:P59535}; Taste receptor type 2 member 18; Taste receptor type 2 member 40; Taste receptor type 2 member 58; taste receptor, type 2, member 40
Gene Aliases: GPR60; mt2r33; RGD1562314; T2r18; T2r33; T2R40; T2R58; Tas2r144; TAS2R40; Tas2r44
UniProt ID: (Human) P59535, (Rat) Q67ET2, (Mouse) Q7TQB8
Entrez Gene ID: (Human) 259286, (Rat) 500101, (Mouse) 387515
Molecular Function: transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support