Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
The antibody generated using this antigen is expected to also recognise TAS2R44, TAS2R45 and TAS2R47.
Specificity of this antibody: TAS2R43 (and also TAS2R44, TAS2R45 and TAS2R47. See the Immunogen).
TAS2R43 belongs to the large TAS2R receptor family. TAS2Rs are expressed on the surface of taste receptor cells and mediate the perception of bitterness through a G protein-coupled second messenger pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: T2R43; T2R52; Taste receptor type 2 member 43; Taste receptor type 2 member 52; taste receptor, type 2, member 43
Gene Aliases: T2R43; T2R52; TAS2R43
UniProt ID: (Human) P59537
Entrez Gene ID: (Human) 259289
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support