Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MMSFLHIVFS ILVVVAFILG NFANGFIALI NFIAWVKRQK ISSADQIIAA LAVSRVGLLW VILLHWYSTV LNPTSSNLKV IIFISNAWAV TNHFSIWLAT SLSIFYLLKI VNFSRLIFHH LKRKAKSVVL VIVLGSLFFL ICQLVMKNTY INVWTEECEG NVTWKIKLRN AMHLSNLTVA MLANLIPFTL TLISFLLLIY SLCKHLKKMQ LHGKGSQDPS TKIHIKALQT VTSFLVLLAI YFLCLIISFW NSKMLPKEIV LMLCQAFGII YPSFHSFILI WGNKTLKQTF LSVLWQVTCW AKGQNQSTP
The sense of taste provides animals with valuable information about the quality and nutritional value of food. A family of G protein-coupled receptors are involved in taste perception and include T1R, which is involved in sweet and umami taste perception and T2R, which is involved in bitter taste perception. Both types of taste receptors couple to various G proteins to initiate signal transduction cascades. T2R49 plays a role in sensing the chemical composition of the gastrointestinal content. T2R49 is expressed in subsets of taste receptor cells of the tongue and exclusively in gustducin-positive cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC119118; MGC119119; MGC119120; MGC119121; MGC134637; T2R49; T2R56; Taste receptor type 2 member 20; Taste receptor type 2 member 49; Taste receptor type 2 member 56; taste receptor, type 2, member 20; taste receptor, type 2, member 49
Gene Aliases: T2R20; T2R49; T2R56; TAS2R20; TAS2R49
UniProt ID: (Human) P59543
Entrez Gene ID: (Human) 259295
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support