Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MTMEKGMSSG EGLPSRSSQV SAGKITAKEL ETKQSYKEKR GGFVLVHAGA GYHSESKAKE YKHVCKRACQ KAIEKLQAGA LATDAVTAAL VELEDSPFTN AGMGSNLNLL GEIECDASIM DGKSLNFGAV GALSGIKNPV SVANRLLCEG QKGKLSAGRI PPCFLVGEGA YRWAVDHGIP SCPPNIMTTR FSLAAFKRNK RKLELAERVD TDFMQLKKRR QSSEKENDSG TLDTVGAVVV DHEGNVAAAV SSGGLALKHP GRVGQAALYG CGCWAENTGA HNPYSTAVST SGCGEHLVRT ILARECSHAL QAEDAHQALL ETMQNKFISS PFLASEDGVL GGVIVLRSCR CSAEPDSSQN KQTLLVEFLW SHTTESMCVG YMSAQDGKAK THISRLPPGA VAGQSVAIEG GVCRLESPVN
This gene encodes an endopeptidase that cleaves specific substrates following aspartate residues. The encoded protein undergoes posttranslational autoproteolytic processing to generate alpha and beta subunits, which reassemble into the active alpha2-beta2 heterotetramer. It is required to cleave MLL, a protein required for the maintenance of HOX gene expression, and TFIIA, a basal transcription factor. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20212; MGC39159; Taspase-1; Threonine aspartase 1
Gene Aliases: C20orf13; dJ585I14.2; TASP1
UniProt ID: (Human) Q9H6P5
Entrez Gene ID: (Human) 55617
Molecular Function:
protease
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support