Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNPASAPPP LPPPGQQVIH VTQDLDTDLE ALFNSVMNPK PSSWRKKILP ESFFKEPDSG SHSRQSSTDS SGGHPGPRLA GGAQHVRSHS SPASLQLGTG AGAAGSPAQQ HAHLRQQSYD VTDELPLPPG WEMTFTATGQ RYFLNHIEKI TTWQDPRKAM NQPLNHMNLH PAVSSTPVPQ RSMAVSQPNL VMNHQHQQQM APSTLSQQNH PTQNPPAGLM SMPNALTTQQ QQQQKLRLQR IQMERERIRM RQEELMRQEA ALCRQLPMEA ETLAPVQAAV NPPTMTPDMR SITNNSSDPF LNGGPYHSRE QSTDSGLGLG CYSVPTTPED FLSNVDEMDT GENAGQTPMN INPQQTRFPD FLDCLPGTNV DLGTLESEDL IPLFNDVESA LNKSEPFLTW L (1-400 aa encoded by BC014052)
WW domain-containing transcription regulator protein 1 (WWTR1) is a transcriptional coactivator which acts as a downstream regulatory target in the Hippo signaling pathway that plays a pivotal role in organ size control and tumor supression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1. In turn, MOB1 phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. WWTR1 enhances PAX8 and NKX2-1/TTF1-dependent gene activation. WWTR1 regulates the nuclear accumulation of SMADS and has a key role in coupling them to the transcriptional machinery such as the mediator complex. WWTR1 also regulates embryonic stem-cell self-renewal and promotes cell proliferation and epithelial-mesenchymal transition.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp586I1419; FLJ27004; FLJ45718; transcriptional co-activator with PDZ-binding motif; transcriptional co-activator with PDZ-binding motif (TA)Z; transcriptional co-activator with PDZ-binding motif (TAZ); transcriptional coactivator with PDZ binding motif; Transcriptional coactivator with PDZ-binding motif; WW domain-containing transcription regulator protein 1
Gene Aliases: 2310058J06Rik; 2610021I22Rik; C78399; TAZ; WWTR1
UniProt ID: (Human) Q9GZV5, (Mouse) Q9EPK5
Entrez Gene ID: (Human) 25937, (Mouse) 97064, (Rat) 295062
Molecular Function: transcription cofactor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support