Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPITFTARK HLLSNEVSVD FGLQLVGSLP VHSLTTMPML PWVVAEVRRL SRQSTRKEPV TKQVRLCVSP SGLRCEPEPG RSQQWDPLIY SSIFECKPQR VHKLIHNSHD PSYFACLIKE DAVHRQSICY VFKADDQTKV PEIISSIRQA GKIARQEELH CPSEFDDTFS KKFEVLFCGR VTVAHKKAPP ALIDECIEKF NHVSGSRGSE SPRPNPPHAA PTGSQEPVRR PMRKSFSQPG LRSLAFRKEL QDGGLRSSGF FSSFEESDIE NHLISGHNIV QPTDIEENRT MLFTIGQSEV YLISPDTKKI ALEKNFKEIS FCSQGIRHVD HFGFICRESS GGGGFHFVCY VFQCTNEALV DEIMMTLKQA FTVAAVQQTA KAPAQLCEGC PLQSLHKLCE RIEGMNSSKT KLELQKHLTT LTNQEQATIF EEVQKLRPRN EQRENELIIS FLRCLYEEKQ KEHIHIGEMK QTSQMAAENI GSELPPSATR FRLDMLKNKA KRSLTESLES ILSRGNKARG LQEHSISVDL DSSLSSTLSN TSKEPSVCEK EALPISESSF KLLGSSEDLS SDSESHLPEE PAPLSPQQAF RRRANTLSHF PIECQEPPQP ARGSPGVSQR KLMRYHSVST ETPHERKDFE SKANHLGDSG GTPVKTRRHS WRQQIFLRVA TPQKACDSSS RYEDYSELGE LPPRSPLEPV CEDGPFGPPP EEKKRTSREL RELWQKAILK QILLLRMEKE NQKLQASEND LLNKRLKLDY EEITPCLKEV TTVWEKMLST PGRSKIKFDM EKMHSAVGQG VPRHHRGEIW KFLAEQFHLK HQFPSKQQPK DVPYKELLKQ LTSQQHAILI DLGRTFPTHP YFSAQLGAGQ LSLYNILKAY SLLDQEVGYC QGLSFVAGIL LLHMSEEEAF KMLKFLMFDM GLRKQYRPDM IILQIQMYQL SRLLHDYHRD LYNHLEEHEI GPSLYAAPWF LTMFASQFPL GFVARVFDMI FLQGTEVIFK VALSLLGSHK PLILQHENLE TIVDFIKSTL PNLGLVQMEK TINQVFEMDI AKQLQAYEVE YHVLQEELID SSPLSDNQRM DKLEKTNSSL RKQNLDLLEQ LQVANGRIQS LEATIEKLLS SESKLKQAML TLELERSALL QTVEELRRRS AEPSDREPEC TQPEPTGD
The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1; TBC1 domain family member 1
Gene Aliases: KIAA1108; TBC; TBC1; TBC1D1
UniProt ID: (Human) Q86TI0
Entrez Gene ID: (Human) 23216
Molecular Function:
scaffold/adaptor protein
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support