Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSLHKSRIA DFQDVLKEPS IALEKLRELS FSGIPCEGGL RCLCWKILLN YLPLERASWT SILAKQRELY AQFLREMIIQ PGIAKANMGV SREDVTFEDH PLNPNPDSRW NTYFKDNEVL LQIDKDVRRL CPDISFFQRA TDYPCLLILD PQNEFETLRK RVEQTTLKSQ TVARNRSGVT NMSSPHKNSV PSSLNEYEVL PNGCEAHWEV VERILFIYAK LNPGIAYVQG MNEIVGPLYY TFATDPNSEW KEHAEADTFF CFTNLMAEIR DNFIKSLDDS QCGITYKMEK VYSTLKDKDV ELYLKLQEQN IKPQFFAFRW LTLLLSQEFL LPDVIRIWDS LFADDNRFDF LLLVCCAMLM LIREQLLEGD FTVNMRLLQD YPITDVCQIL QKAKELQDSK
TBC1D13 may act as a GTPase-activating protein for Rab family protein(s).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TBC1 domain family member 13; TBC1 domain family, member 13
Gene Aliases: TBC1D13
UniProt ID: (Human) Q9NVG8
Entrez Gene ID: (Human) 54662
Molecular Function:
GTPase-activating protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support